Product Details
- SNP ID
-
rs202087879
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:16680015 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGAACTGTCCCGGCGCCACCCCAT[C/T]GCGTCCTGGCTGTGCGCCATGCTGC
- Phenotype
-
MIM: 611235
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM38A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs730120] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TMEM38A
- Gene Name
- transmembrane protein 38A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024074.2 |
247 |
Silent Mutation |
ATC,ATT |
I52I |
NP_076979.1 |
View Full Product Details