Product Details

SNP ID
rs200738242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55230639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGCCTGGGGATGCAGGCCCTTC[C/T]GGGGCAGAGCCATTGGGTATCGGAG
Phenotype
MIM: 610875
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR6802 PubMed Links

Gene Details

Gene
MIR6802
Gene Name
microRNA 6802
There are no transcripts associated with this gene.

Gene
MIR6804
Gene Name
microRNA 6804
There are no transcripts associated with this gene.

Gene
PPP6R1
Gene Name
protein phosphatase 6 regulatory subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014931.3 2858 Silent Mutation CCA,CCG P872P NP_055746.3
Gene
TMEM86B
Gene Name
transmembrane protein 86B
There are no transcripts associated with this gene.

View Full Product Details