Product Details
- SNP ID
-
rs201359702
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:13954554 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATCTTCCTGGGCTTTTCCAAATGC[A/G]GCCGCTACGTCCTCTCCTACACCAG
- Phenotype
-
MIM: 600006
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DCAF15
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10409785] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DCAF15
- Gene Name
- DDB1 and CUL4 associated factor 15
There are no transcripts associated with this gene.
- Gene
- PODNL1
- Gene Name
- podocan like 1
There are no transcripts associated with this gene.
- Gene
- RFX1
- Gene Name
- regulatory factor X1
There are no transcripts associated with this gene.
View Full Product Details