Product Details

SNP ID
rs201735546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36013025 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTAGGGCACTGAGGTCACCTGT[C/G]GAAGCAAATACTCCTCCTCTTCTTT
Phenotype
MIM: 613304 MIM: 607382 MIM: 615535
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ALKBH6 PubMed Links

Gene Details

Gene
ALKBH6
Gene Name
alkB homolog 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297701.1 256 Missense Mutation CCA,CGA P40R NP_001284630.1
NM_032878.3 256 Missense Mutation CCA,CGA P68R NP_116267.3
NM_198867.1 256 Missense Mutation CCA,CGA P68R NP_942567.1
XM_005259357.4 256 Missense Mutation CCA,CGA P40R XP_005259414.1
Gene
CLIP3
Gene Name
CAP-Gly domain containing linker protein 3
There are no transcripts associated with this gene.

Gene
LOC101927572
Gene Name
uncharacterized LOC101927572
There are no transcripts associated with this gene.

Gene
SYNE4
Gene Name
spectrin repeat containing nuclear envelope family member 4
There are no transcripts associated with this gene.

View Full Product Details