Product Details
- SNP ID
-
rs202195204
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:47066417 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTAAAAACATCCTTCAGGGATGCC[A/G]CATCCTGCTCACCCTCGCGGGCCGG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZC3H4
PubMed Links
Gene Details
- Gene
- ZC3H4
- Gene Name
- zinc finger CCCH-type containing 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015168.1 |
5164 |
Missense Mutation |
GCG,GTG |
A1284V |
NP_055983.1 |
XM_005258676.3 |
5164 |
Missense Mutation |
GCG,GTG |
A1248V |
XP_005258733.1 |
XM_005258677.4 |
5164 |
Missense Mutation |
GCG,GTG |
A1243V |
XP_005258734.1 |
XM_005258678.2 |
5164 |
Missense Mutation |
GCG,GTG |
A1235V |
XP_005258735.1 |
XM_006723113.3 |
5164 |
Missense Mutation |
GCG,GTG |
A1284V |
XP_006723176.1 |
XM_011526668.2 |
5164 |
Missense Mutation |
GCG,GTG |
A1243V |
XP_011524970.1 |
XM_011526669.2 |
5164 |
Missense Mutation |
GCG,GTG |
A1238V |
XP_011524971.1 |
XM_011526670.2 |
5164 |
Missense Mutation |
GCG,GTG |
A1234V |
XP_011524972.1 |
XM_017026529.1 |
5164 |
Missense Mutation |
GCG,GTG |
A1271V |
XP_016882018.1 |
XM_017026530.1 |
5164 |
Missense Mutation |
GCG,GTG |
A1200V |
XP_016882019.1 |
XM_017026531.1 |
5164 |
Missense Mutation |
GCG,GTG |
A987V |
XP_016882020.1 |
View Full Product Details