Product Details

SNP ID
rs200808399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55529920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGAACCAGGGCTGAGGGCGGTCC[C/T]GGGGCTGGCCCGACGCCTGCCAGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SBK2 PubMed Links

Gene Details

Gene
SBK2
Gene Name
SH3 domain binding kinase family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101401.2 875 Missense Mutation CAG,CGG Q287R NP_001094871.2
XM_006723327.3 875 Missense Mutation CAG,CGG Q287R XP_006723390.1
XM_011527227.2 875 Intron XP_011525529.1

View Full Product Details