Product Details

SNP ID
rs201117247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38320117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCAGCGCAGCCCGTGTGTGGCTG[C/G]CCCCGCCCTGGACGCCTTCGTGGAG
Phenotype
MIM: 603939
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KCNK6 PubMed Links

Gene Details

Gene
KCNK6
Gene Name
potassium two pore domain channel subfamily K member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004823.1 251 Missense Mutation GCC,GGC A56G NP_004814.1
XM_011527526.1 251 UTR 5 XP_011525828.1
XM_011527527.1 251 Missense Mutation GCC,GGC A56G XP_011525829.1
XM_017027506.1 251 Missense Mutation GCC,GGC A56G XP_016882995.1
Gene
YIF1B
Gene Name
Yip1 interacting factor homolog B, membrane trafficking protein
There are no transcripts associated with this gene.

View Full Product Details