Product Details

SNP ID
rs201703893
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8490623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTTGGGGTCCCGGCACAGCATC[A/T]CCTCATTGCTGGTGAACTCGATCAC
Phenotype
MIM: 160994 MIM: 606466
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
HNRNPM PubMed Links

Gene Details

Gene
HNRNPM
Gene Name
heterogeneous nuclear ribonucleoprotein M
There are no transcripts associated with this gene.

Gene
PRAM1
Gene Name
PML-RARA regulated adaptor molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032152.4 1946 Missense Mutation GAG,GTG E626V NP_115528.4
XM_005272502.2 1946 Missense Mutation GAG,GTG E626V XP_005272559.1
XM_011528352.2 1946 Missense Mutation GAG,GTG E628V XP_011526654.1
XM_011528353.2 1946 Missense Mutation GAG,GTG E628V XP_011526655.1

View Full Product Details