Product Details
- SNP ID
-
rs200982088
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:38352363 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCACCGAGAGCACCCTCTTCATT[C/T]GGCAGAACCAGCTGGTCTACTATTT
- Phenotype
-
MIM: 613452
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CATSPERG
PubMed Links
Gene Details
- Gene
- CATSPERG
- Gene Name
- cation channel sperm associated auxiliary subunit gamma
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021185.4 |
1631 |
Missense Mutation |
CGG,TGG |
R310W |
NP_067008.3 |
XM_005259114.4 |
1631 |
Missense Mutation |
CGG,TGG |
R310W |
XP_005259171.1 |
XM_005259115.4 |
1631 |
Missense Mutation |
CGG,TGG |
R310W |
XP_005259172.1 |
XM_005259116.2 |
1631 |
Missense Mutation |
CGG,TGG |
R310W |
XP_005259173.1 |
XM_006723308.3 |
1631 |
Missense Mutation |
CGG,TGG |
R310W |
XP_006723371.1 |
XM_006723310.2 |
1631 |
Missense Mutation |
CGG,TGG |
R310W |
XP_006723373.1 |
XM_006723311.2 |
1631 |
Missense Mutation |
CGG,TGG |
R310W |
XP_006723374.1 |
XM_011527174.2 |
1631 |
Missense Mutation |
CGG,TGG |
R310W |
XP_011525476.1 |
XM_011527175.2 |
1631 |
Missense Mutation |
CGG,TGG |
R287W |
XP_011525477.1 |
XM_011527176.2 |
1631 |
Missense Mutation |
CGG,TGG |
R310W |
XP_011525478.1 |
XM_011527177.2 |
1631 |
Missense Mutation |
CGG,TGG |
R270W |
XP_011525479.1 |
XM_011527178.2 |
1631 |
Missense Mutation |
CGG,TGG |
R195W |
XP_011525480.1 |
XM_011527179.1 |
1631 |
Missense Mutation |
CGG,TGG |
R310W |
XP_011525481.1 |
XM_011527182.2 |
1631 |
Intron |
|
|
XP_011525484.1 |
XM_017027053.1 |
1631 |
Missense Mutation |
CGG,TGG |
R310W |
XP_016882542.1 |
XM_017027054.1 |
1631 |
Missense Mutation |
CGG,TGG |
R195W |
XP_016882543.1 |
XM_017027055.1 |
1631 |
Intron |
|
|
XP_016882544.1 |
View Full Product Details