Product Details

SNP ID
rs201222388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1387815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCGTGGTCCTCTCTGCAGCTCC[C/T]GGCCTGCGCGGCTTCCGGATCCTTG
Phenotype
MIM: 601240 MIM: 601825
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAMT PubMed Links

Gene Details

Gene
GAMT
Gene Name
guanidinoacetate N-methyltransferase
There are no transcripts associated with this gene.

Gene
MUM1
Gene Name
melanoma associated antigen (mutated) 1
There are no transcripts associated with this gene.

Gene
NDUFS7
Gene Name
NADH:ubiquinone oxidoreductase core subunit S7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024407.4 64 Silent Mutation CCC,CCT P7P NP_077718.3
XM_005259556.4 64 Silent Mutation CCC,CCT P7P XP_005259613.2
XM_017026768.1 64 Silent Mutation CCC,CCT P92P XP_016882257.1

View Full Product Details