Product Details

SNP ID
rs201533373
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35778506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCGGAGTTACGATGACTTTCGT[C/T]CCCTGGATGCCCACCTCCACCGGTG
Phenotype
MIM: 614902
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP33 PubMed Links

Gene Details

Gene
ARHGAP33
Gene Name
Rho GTPase activating protein 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172630.1 1853 UTR 5 NP_001166101.1
NM_052948.3 1853 Missense Mutation CCC,TCC P105S NP_443180.2
XM_005258488.1 1853 Missense Mutation CCC,TCC P105S XP_005258545.1
XM_006722999.1 1853 Missense Mutation CCC,TCC P105S XP_006723062.1
XM_006723003.2 1853 Intron XP_006723066.1
XM_011526416.1 1853 UTR 5 XP_011524718.1
XM_011526417.1 1853 UTR 5 XP_011524719.1
XM_011526418.1 1853 UTR 5 XP_011524720.1
XM_011526419.1 1853 UTR 5 XP_011524721.1
XM_011526420.1 1853 UTR 5 XP_011524722.1
XM_011526421.2 1853 Missense Mutation CCC,TCC P142S XP_011524723.1
XM_017026238.1 1853 Missense Mutation CCC,TCC P142S XP_016881727.1
Gene
PROSER3
Gene Name
proline and serine rich 3
There are no transcripts associated with this gene.

View Full Product Details