Product Details

SNP ID
rs200641863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12351063 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCATAGCGTTTCTTTCCAGTGTG[A/G]GGTCTTTCCTGTGTTTGAAAGGAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF442 PubMed Links

Gene Details

Gene
ZNF442
Gene Name
zinc finger protein 442
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030824.2 561 Silent Mutation CCC,CCT P174P NP_110451.1
XM_006722908.3 561 Silent Mutation CCC,CCT P167P XP_006722971.1
XM_017027316.1 561 Silent Mutation CCC,CCT P182P XP_016882805.1
XM_017027317.1 561 Silent Mutation CCC,CCT P117P XP_016882806.1
XM_017027318.1 561 Silent Mutation CCC,CCT P174P XP_016882807.1
Gene
ZNF563
Gene Name
zinc finger protein 563
There are no transcripts associated with this gene.

View Full Product Details