Product Details

SNP ID
rs201155665
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45795938 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCTCCTCTGTGGCTCCCAGGGC[A/T]TGTTCCTGGGCTGCTTTCAGAGCCT
Phenotype
MIM: 609857 MIM: 607548
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DMWD PubMed Links

Gene Details

Gene
DMWD
Gene Name
dystrophia myotonica, WD repeat containing
There are no transcripts associated with this gene.

Gene
RSPH6A
Gene Name
radial spoke head 6 homolog A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030785.3 2256 Missense Mutation CAA,CAT Q695H NP_110412.1
XM_011527351.1 2256 Intron XP_011525653.1

View Full Product Details