Product Details

SNP ID
rs201704776
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:1918024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTCCTTCCTCTCTGGGCTTGGCC[A/G]TCTTGACAGCTTCATCGTAGGAGGG
Phenotype
MIM: 610171
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CALML6 PubMed Links

Gene Details

Gene
CALML6
Gene Name
calmodulin like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005244729.3 524 Intron XP_005244786.2
XM_011540841.2 524 Intron XP_011539143.1
Gene
CFAP74
Gene Name
cilia and flagella associated protein 74
There are no transcripts associated with this gene.

Gene
TMEM52
Gene Name
transmembrane protein 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178545.3 524 Missense Mutation ACG,ATG T163M NP_848640.1

View Full Product Details