Product Details
- SNP ID
-
rs201361420
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
11
- Location
-
Chr.1:232805678 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTTTACCCTCCTGACGGCCCCGGC[A/G]CTCCCGCCGCCGAACCGTGGCCCGG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MAP10
PubMed Links
Gene Details
- Gene
- MAP10
- Gene Name
- microtubule associated protein 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_019090.2 |
787 |
Missense Mutation |
ACT,GCT |
T219A |
NP_061963.2 |
View Full Product Details