Product Details

SNP ID
rs201123337
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:35716094 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGCTGCGCACCCCTTCCTCAGG[G/T]GATCCAGGGGCCTGGAAGGCTTGGT
Phenotype
MIM: 605434
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C1orf216 PubMed Links

Gene Details

Gene
C1orf216
Gene Name
chromosome 1 open reading frame 216
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152374.1 716 Silent Mutation TCA,TCC S76S NP_689587.1
Gene
CLSPN
Gene Name
claspin
There are no transcripts associated with this gene.

View Full Product Details