Product Details

SNP ID
rs201362280
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:223393952 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGAGCTGGCACTCCCCTGAGTGG[A/C]ACATTCAGGGTAGAAAAGGCACAGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCDC185 PubMed Links

Gene Details

Gene
CCDC185
Gene Name
coiled-coil domain containing 185
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152610.2 580 Silent Mutation GGA,GGC G159G NP_689823.2

View Full Product Details