Product Details

SNP ID
rs201494815
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:109507497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTGGATCCGACATCCTCCGTTG[A/G]CCCTTGCCTGTGGCCTCAGGCACTG
Phenotype
MIM: 615689
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AMIGO1 PubMed Links

Gene Details

Gene
AMIGO1
Gene Name
adhesion molecule with Ig like domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020703.2 1804 Silent Mutation GGC,GGT G472G NP_065754.2
XM_011541812.2 1804 Silent Mutation GGC,GGT G472G XP_011540114.1
Gene
CYB561D1
Gene Name
cytochrome b561 family member D1
There are no transcripts associated with this gene.

View Full Product Details