Product Details

SNP ID
rs201560600
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:20721946 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGAAGAGGCTACGTTCAGTAACC[A/G]CGTGGTTCTCCACCAGAGCCTCCAG
Phenotype
MIM: 605037
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF17 PubMed Links

Gene Details

Gene
KIF17
Gene Name
kinesin family member 17
There are no transcripts associated with this gene.

Gene
SH2D5
Gene Name
SH2 domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001103160.1 1198 Missense Mutation GCG,GTG A289V NP_001096630.1
NM_001103161.1 1198 Missense Mutation GCG,GTG A373V NP_001096631.1
XM_011541459.1 1198 Missense Mutation GCG,GTG A373V XP_011539761.1
XM_011541460.1 1198 Missense Mutation GCG,GTG A373V XP_011539762.1
XM_011541461.2 1198 Missense Mutation GCG,GTG A373V XP_011539763.1
XM_011541462.1 1198 Missense Mutation GCG,GTG A372V XP_011539764.1

View Full Product Details