Product Details

SNP ID
rs201679731
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:197088321 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACAAGTTTTTGTAAAAATGCTTC[C/T]GCCTTTGTCTGCAACTTTATTACCA
Phenotype
MIM: 605481
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASPM PubMed Links

Gene Details

Gene
ASPM
Gene Name
abnormal spindle microtubule assembly
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206846.1 10353 Missense Mutation AGA,GGA R1781G NP_001193775.1
NM_018136.4 10353 Missense Mutation AGA,GGA R3366G NP_060606.3

View Full Product Details