Product Details

SNP ID
rs201005880
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:241500535 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGATTCCCACCACGCAGTTTTCT[A/G]TAAAGGAAACTGAAGCATCCCCCAG
Phenotype
MIM: 136850
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FH PubMed Links

Gene Details

Gene
FH
Gene Name
fumarate hydratase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000143.3 1795 Missense Mutation ACA,ATA T431I NP_000134.2
XM_011544132.2 1795 Missense Mutation ACA,ATA T355I XP_011542434.1

View Full Product Details