Product Details

SNP ID
rs201336358
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:75918974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTTAGATTTCTAAATTCAGATAG[A/T]TTTGTAGACGAGCAGGAATTAGAAG
Phenotype
MIM: 602105
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ASB17 PubMed Links

Gene Details

Gene
ASB17
Gene Name
ankyrin repeat and SOCS box containing 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080868.2 1006 Missense Mutation AAC,ATC N289I NP_543144.1
XM_006710345.3 1006 Missense Mutation AAC,ATC N241I XP_006710408.1
XM_006710346.3 1006 Missense Mutation AAC,ATC N211I XP_006710409.1
Gene
MSH4
Gene Name
mutS homolog 4
There are no transcripts associated with this gene.

View Full Product Details