Product Details

SNP ID
rs201534275
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:20143528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGAGGAGCGCCATCCTGTCCTT[C/T]GTGGGCTACGGTTGCTACTGTGGGC
Phenotype
MIM: 616793
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLA2G2F PubMed Links

Gene Details

Gene
PLA2G2F
Gene Name
phospholipase A2 group IIF
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022819.3 343 Silent Mutation TTC,TTT F84F NP_073730.3
XM_011541955.1 343 Silent Mutation TTC,TTT F84F XP_011540257.1
XM_011541956.1 343 Silent Mutation TTC,TTT F84F XP_011540258.1

View Full Product Details