Product Details

SNP ID
rs201284937
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:170532166 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACCCGGATGAGCTGGGCAGCAGT[A/G]TTGGCAGTCGCGGCTGCGAGATTTG
Phenotype
MIM: 607983
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GORAB PubMed Links

Gene Details

Gene
GORAB
Gene Name
golgin, RAB6 interacting
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146039.1 45 Silent Mutation GTA,GTG V6V NP_001139511.1
NM_001320252.1 45 UTR 5 NP_001307181.1
NM_152281.2 45 Silent Mutation GTA,GTG V6V NP_689494.2
XM_006711628.3 45 UTR 5 XP_006711691.1
XM_011510149.2 45 Silent Mutation GTA,GTG V6V XP_011508451.1
XM_011510150.2 45 UTR 5 XP_011508452.1
XM_011510151.1 45 UTR 5 XP_011508453.1
XM_017002807.1 45 UTR 5 XP_016858296.1
Gene
LOC101928650
Gene Name
uncharacterized LOC101928650
There are no transcripts associated with this gene.

View Full Product Details