Product Details

SNP ID
rs201682049
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:109075933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGGTCACTCACCGTGGGGTCTTC[C/T]TCCTCATCTGCCATCCCACTAGCAC
Phenotype
MIM: 600774
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAF13 PubMed Links

Gene Details

Gene
TAF13
Gene Name
TATA-box binding protein associated factor 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005645.3 70 Silent Mutation GAA,GAG E5E NP_005636.1

View Full Product Details