Product Details

SNP ID
rs201099099
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:248406311 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATCCAAACAGATTTGCGCCTTC[A/G]TACACCCATGTACTTCCTCCTCAGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2T1 PubMed Links

Gene Details

Gene
OR2T1
Gene Name
olfactory receptor family 2 subfamily T member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030904.1 317 Missense Mutation CAT,CGT H106R NP_112166.1

View Full Product Details