Product Details

SNP ID
rs201060319
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:247814969 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGAAGCTGGCTTCAGATAAGCA[A/G]TGAATCCAGTGGAAAGAAATAACAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR14A16 PubMed Links

Gene Details

Gene
OR14A16
Gene Name
olfactory receptor family 14 subfamily A member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001966.1 761 Missense Mutation ACT,ATT T254I NP_001001966.1

View Full Product Details