Product Details

SNP ID
rs201220815
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:6581240 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGACAAGAAACCCGAGGACTGC[A/G]AAGTGCCCCCAAGGCCCTTAGAGGC
Phenotype
MIM: 616262 MIM: 606225 MIM: 165270
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHL21 PubMed Links

Gene Details

Gene
KLHL21
Gene Name
kelch like family member 21
There are no transcripts associated with this gene.

Gene
TAS1R1
Gene Name
taste 1 receptor member 1
There are no transcripts associated with this gene.

Gene
ZBTB48
Gene Name
zinc finger and BTB domain containing 48
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278647.1 780 Missense Mutation AAA,GAA K211E NP_001265576.1
NM_001278648.1 780 Missense Mutation AAA,GAA K211E NP_001265577.1
NM_005341.3 780 Missense Mutation AAA,GAA K211E NP_005332.1
XM_017001110.1 780 Missense Mutation AAA,GAA K211E XP_016856599.1
XM_017001111.1 780 Missense Mutation AAA,GAA K211E XP_016856600.1
XM_017001112.1 780 UTR 5 XP_016856601.1
XM_017001113.1 780 UTR 5 XP_016856602.1
XM_017001114.1 780 UTR 5 XP_016856603.1

View Full Product Details