Product Details

SNP ID
rs201221009
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:86469017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTTCATCTTGATTCTTCACCTT[C/T]TAGAAGGGGCCCTGAGTAATTCACT
Phenotype
MIM: 603906
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLCA1 PubMed Links

Gene Details

Gene
CLCA1
Gene Name
chloride channel accessory 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285.3 175 Silent Mutation CTA,TTA L16L NP_001276.2

View Full Product Details