Product Details

SNP ID
rs201537248
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:23431737 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTACATCTCCGAAGGGGTGGTCCC[A/G]GATGTCAGGCTGGGTCTGGAGGTGC
Phenotype
MIM: 616594 MIM: 604128
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASAP3 PubMed Links

Gene Details

Gene
ASAP3
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143778.1 2629 Silent Mutation TCC,TCT S826S NP_001137250.1
NM_017707.3 2629 Silent Mutation TCC,TCT S835S NP_060177.2
XM_011541755.1 2629 Silent Mutation TCC,TCT S858S XP_011540057.1
XM_017001685.1 2629 Silent Mutation TCC,TCT S856S XP_016857174.1
XM_017001686.1 2629 Silent Mutation TCC,TCT S837S XP_016857175.1
XM_017001687.1 2629 Silent Mutation TCC,TCT S815S XP_016857176.1
XM_017001688.1 2629 Silent Mutation TCC,TCT S725S XP_016857177.1
XM_017001689.1 2629 Silent Mutation TCC,TCT S706S XP_016857178.1
Gene
TCEA3
Gene Name
transcription elongation factor A3
There are no transcripts associated with this gene.

View Full Product Details