Product Details

SNP ID
rs201749906
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:158181684 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTTTATCGAAGCAGCTTCACCAG[A/G]GACGTGAAGGAATTCGCCAAAATGC
Phenotype
MIM: 188410
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD1D PubMed Links

Gene Details

Gene
CD1D
Gene Name
CD1d molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319145.1 469 Silent Mutation AGA,AGG R97R NP_001306074.1
NM_001766.3 469 Silent Mutation AGA,AGG R97R NP_001757.1
XM_005245583.1 469 UTR 5 XP_005245640.1
XM_006711621.1 469 UTR 5 XP_006711684.1
XM_011510125.1 469 Silent Mutation AGA,AGG R97R XP_011508427.1
XM_011510127.1 469 UTR 5 XP_011508429.1

View Full Product Details