Product Details

SNP ID
rs201766470
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:13583910 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCTCCGGGGCTCCTGCTCCCAC[A/C]CCTCCGGCCCCCCCACCGTCGCGCT
Phenotype
MIM: 608863
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PDPN PubMed Links

Gene Details

Gene
PDPN
Gene Name
podoplanin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006624.1 154 Intron NP_001006625.1
NM_001006625.1 154 Intron NP_001006626.1
NM_006474.4 154 Silent Mutation ACA,ACC T35T NP_006465.3
NM_198389.2 154 Silent Mutation ACA,ACC T35T NP_938203.2
XM_006710295.1 154 Intron XP_006710358.1

View Full Product Details