Product Details

SNP ID
rs201768904
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:167810846 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCATTAATATACAGACGTAAGCC[A/G]TCAGGTGGTAGAGCCTTGGGCAAAA
Phenotype
MIM: 605205
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADCY10 PubMed Links

Gene Details

Gene
ADCY10
Gene Name
adenylate cyclase 10, soluble
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167749.2 6027 Missense Mutation ACG,ATG T1364M NP_001161221.1
NM_001297772.1 6027 Missense Mutation ACG,ATG T1425M NP_001284701.1
NM_018417.5 6027 Missense Mutation ACG,ATG T1517M NP_060887.2
XM_005245330.4 6027 Intron XP_005245387.1
XM_006711449.3 6027 Intron XP_006711512.1
XM_011509760.2 6027 Missense Mutation ACG,ATG T1517M XP_011508062.1
XM_011509762.2 6027 Missense Mutation ACG,ATG T1469M XP_011508064.1
XM_011509763.2 6027 Missense Mutation ACG,ATG T1463M XP_011508065.1
XM_011509766.2 6027 Intron XP_011508068.1
XM_017001778.1 6027 Intron XP_016857267.1

View Full Product Details