Product Details

SNP ID
rs201221908
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:39525764 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAACGTCATCCTGCGCAAGCACC[A/G]CAACATGGTGGTCAAGGCCTGCGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BMP8A PubMed Links

Gene Details

Gene
BMP8A
Gene Name
bone morphogenetic protein 8a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181809.3 1521 Missense Mutation CAC,CGC H392R NP_861525.2
XM_006710616.3 1521 Intron XP_006710679.1
XM_011541381.2 1521 Intron XP_011539683.1
XM_011541382.2 1521 Intron XP_011539684.1
XM_017001198.1 1521 Intron XP_016856687.1
Gene
OXCT2P1
Gene Name
3-oxoacid CoA-transferase 2 pseudogene 1
There are no transcripts associated with this gene.

Gene
PPIEL
Gene Name
peptidylprolyl isomerase E like pseudogene
There are no transcripts associated with this gene.

View Full Product Details