Product Details
- SNP ID
-
rs201498581
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:248349393 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTCTCTGCTCTGGGGGTAGGTGGC[A/G]GCTGTTTCATCTTTATCATCAGGTC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR14C36
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2039824] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR14C36
- Gene Name
- olfactory receptor family 14 subfamily C member 36
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001001918.1 |
619 |
Missense Mutation |
AGC,GGC |
S207G |
NP_001001918.1 |
View Full Product Details