Product Details
- SNP ID
-
rs201524531
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
19
- Location
-
Chr.1:15942545 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCCCGCACCTGTGAGCTGAGTGAC[C/G]GCTGTCGCTGCCAGTGCCTCGGTAG
- Phenotype
-
MIM: 613484
MIM: 604084
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SPEN
PubMed Links
Gene Details
- Gene
- SPEN
- Gene Name
- spen family transcriptional repressor
There are no transcripts associated with this gene.
- Gene
- ZBTB17
- Gene Name
- zinc finger and BTB domain containing 17
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001242884.1 |
2228 |
Silent Mutation |
GCC,GCG |
A592A |
NP_001229813.1 |
NM_001287603.1 |
2228 |
Silent Mutation |
GCC,GCG |
A674A |
NP_001274532.1 |
NM_001287604.1 |
2228 |
Silent Mutation |
GCC,GCG |
A598A |
NP_001274533.1 |
NM_001324137.1 |
2228 |
Silent Mutation |
GCC,GCG |
A598A |
NP_001311066.1 |
NM_001324138.1 |
2228 |
Silent Mutation |
GCC,GCG |
A611A |
NP_001311067.1 |
NM_003443.2 |
2228 |
Silent Mutation |
GCC,GCG |
A674A |
NP_003434.2 |
XM_005245986.1 |
2228 |
Silent Mutation |
GCC,GCG |
A611A |
XP_005246043.1 |
XM_005245987.1 |
2228 |
Silent Mutation |
GCC,GCG |
A598A |
XP_005246044.1 |
XM_011542085.2 |
2228 |
Silent Mutation |
GCC,GCG |
A598A |
XP_011540387.1 |
XM_011542086.1 |
2228 |
Silent Mutation |
GCC,GCG |
A592A |
XP_011540388.1 |
XM_011542087.2 |
2228 |
Silent Mutation |
GCC,GCG |
A592A |
XP_011540389.1 |
XM_011542088.1 |
2228 |
Silent Mutation |
GCC,GCG |
A592A |
XP_011540390.1 |
XM_017002242.1 |
2228 |
Silent Mutation |
GCC,GCG |
A592A |
XP_016857731.1 |
View Full Product Details