Product Details

SNP ID
rs201603810
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:53459636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGGTGCTCAAGACGCAGGCCGC[C/T]GAGGAGGAGCAGGAGGCGGCCCTGT
Phenotype
MIM: 614805
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DMRTB1 PubMed Links

Gene Details

Gene
DMRTB1
Gene Name
DMRT like family B with proline rich C-terminal 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033067.2 238 Silent Mutation GCC,GCT A61A NP_149056.1

View Full Product Details