Product Details

SNP ID
rs201156296
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:113895212 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAATAGCAGTTCTGTTAAGAACAG[A/C]CAGCCAGTATCATCCTGAGCACTGA
Phenotype
MIM: 607245 MIM: 609683
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AP4B1 PubMed Links

Gene Details

Gene
AP4B1
Gene Name
adaptor related protein complex 4 beta 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253852.2 1775 Silent Mutation NP_001240781.1
NM_001253853.2 1775 Silent Mutation NP_001240782.1
NM_001308312.1 1775 Silent Mutation NP_001295241.1
NM_006594.4 1775 Silent Mutation NP_006585.2
XM_011540523.2 1775 Missense Mutation XP_011538825.1
XM_011540524.2 1775 Missense Mutation XP_011538826.1
XM_011540525.2 1775 Missense Mutation XP_011538827.1
XM_011540528.2 1775 Missense Mutation XP_011538830.1
XM_017000088.1 1775 Missense Mutation XP_016855577.1
XM_017000089.1 1775 Missense Mutation XP_016855578.1
XM_017000090.1 1775 Missense Mutation XP_016855579.1
XM_017000091.1 1775 Missense Mutation XP_016855580.1
XM_017000092.1 1775 Missense Mutation XP_016855581.1
XM_017000093.1 1775 Intron XP_016855582.1
Gene
AP4B1-AS1
Gene Name
AP4B1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
BCL2L15
Gene Name
BCL2 like 15
There are no transcripts associated with this gene.

Gene
DCLRE1B
Gene Name
DNA cross-link repair 1B
There are no transcripts associated with this gene.

View Full Product Details