Product Details

SNP ID
rs201169109
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:155659858 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAATCCTTCTTGGCACTGTTTATC[A/G]ACTGGTGGAGGCCCTGGGCTATGTT
Phenotype
MIM: 607860
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
YY1AP1 PubMed Links

Gene Details

Gene
YY1AP1
Gene Name
YY1 associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198899.1 2658 Silent Mutation NP_001185828.1
NM_001198900.1 2658 Silent Mutation NP_001185829.1
NM_001198901.1 2658 Silent Mutation NP_001185830.1
NM_001198902.1 2658 Silent Mutation NP_001185831.1
NM_001198903.1 2658 Silent Mutation NP_001185832.1
NM_001198904.1 2658 Silent Mutation NP_001185833.1
NM_001198905.1 2658 Silent Mutation NP_001185834.1
NM_001198906.1 2658 UTR 3 NP_001185835.1
NM_018253.3 2658 Silent Mutation NP_060723.2
NM_139118.2 2658 Silent Mutation NP_620829.1
NM_139119.2 2658 Silent Mutation NP_620830.1
NM_139121.2 2658 Silent Mutation NP_620832.1

View Full Product Details