Product Details

SNP ID
rs201618193
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:93873123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATTAAGTCACTGTACTTACTCTG[C/T]GTTTTTTCTGAAGTTGATACTTTGA
Phenotype
MIM: 611199
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNTTIP2 PubMed Links

Gene Details

Gene
DNTTIP2
Gene Name
deoxynucleotidyltransferase terminal interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014597.4 1956 Missense Mutation CAC,CGC H633R NP_055412.2

View Full Product Details