Product Details

SNP ID
rs200997544
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:162374048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGACTAACTTTATTGGCATCTCT[A/C]TAGAGACATAAGTAGGCTTCACCGC
Phenotype
MIM: 605551
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C1orf111 PubMed Links

Gene Details

Gene
C1orf111
Gene Name
chromosome 1 open reading frame 111
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011509445.2 Intron XP_011507747.1
Gene
C1orf226
Gene Name
chromosome 1 open reading frame 226
There are no transcripts associated with this gene.

Gene
NOS1AP
Gene Name
nitric oxide synthase 1 adaptor protein
There are no transcripts associated with this gene.

View Full Product Details