Product Details

SNP ID
rs201102562
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:206635728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGGAGGCACAGCTCGTGGGCCT[C/G]GGCGGAAGGATGCGGGGCCGCCTGG
Phenotype
MIM: 603497
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DYRK3 PubMed Links

Gene Details

Gene
DYRK3
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004023.1 193 UTR 5 NP_001004023.1
NM_003582.2 193 Missense Mutation CGG,GGG R9G NP_003573.2
XM_005273315.4 193 Intron XP_005273372.1
XM_011510061.2 193 Intron XP_011508363.1

View Full Product Details