Product Details

SNP ID
rs201116332
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:173041402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTGGGGATTTGCTAGTAAAATGA[C/T]ACCCCAGTATGTATTATTTTTTAGA
Phenotype
MIM: 603898
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNFSF18 PubMed Links

Gene Details

Gene
TNFSF18
Gene Name
tumor necrosis factor superfamily member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005092.3 566 Missense Mutation ATC,GTC I189V NP_005083.2

View Full Product Details