Product Details

SNP ID
rs201183243
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:155256732 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGTGAAGAGCAGGGCGACCAGC[A/G]GCATGAGCACGGATACTGCTGTGTT
Phenotype
MIM: 602989 MIM: 606913
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLK2 PubMed Links

Gene Details

Gene
CLK2
Gene Name
CDC like kinase 2
There are no transcripts associated with this gene.

Gene
FAM189B
Gene Name
family with sequence similarity 189 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267608.1 995 Intron NP_001254537.1
NM_006589.2 995 Intron NP_006580.2
NM_198264.1 995 Intron NP_937995.1
Gene
SCAMP3
Gene Name
secretory carrier membrane protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005698.3 995 Missense Mutation CCG,CTG P280L NP_005689.2
NM_052837.2 995 Missense Mutation CCG,CTG P254L NP_443069.1
XM_006711105.3 995 Missense Mutation CCG,CTG P266L XP_006711168.1
XM_006711106.3 995 Missense Mutation CCG,CTG P213L XP_006711169.1
XM_016999991.1 995 Missense Mutation CGC,TGC R193C XP_016855480.1

View Full Product Details