Product Details

SNP ID
rs201567136
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:93872130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTGTAAAATCTTTTCGGGTCC[A/G]TGCTGGCTCTCATCTTCAGTGCTTT
Phenotype
MIM: 611199
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNTTIP2 PubMed Links

Gene Details

Gene
DNTTIP2
Gene Name
deoxynucleotidyltransferase terminal interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014597.4 2067 Missense Mutation ACG,ATG T670M NP_055412.2

View Full Product Details