Product Details

SNP ID
rs201605916
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:165544661 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCGATCTCCCCATTCACTTTCT[C/T]GGTGCTCAGCAACCTGGTGCAGCTG
Phenotype
MIM: 615218
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC400794 PubMed Links

Gene Details

Gene
LOC400794
Gene Name
uncharacterized LOC400794
There are no transcripts associated with this gene.

Gene
LRRC52
Gene Name
leucine rich repeat containing 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005214.3 421 Missense Mutation TCG,TTG S122L NP_001005214.2

View Full Product Details