Product Details
- SNP ID
-
rs201605987
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
11
- Location
-
Chr.1:42433165 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCTTTGAAGAATTCATCATCAGG[G/T]AGTAAAACATGACCAGGATCTTCAG
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
RIMKLA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1034268] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RIMKLA
- Gene Name
- ribosomal modification protein rimK like family member A
There are no transcripts associated with this gene.
- Gene
- ZMYND12
- Gene Name
- zinc finger MYND-type containing 12
View Full Product Details