Product Details

SNP ID
rs201171352
Assay Type
Functionally Tested
NCBI dbSNP Submissions
4
Location
Chr.1:154569520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGCATCTCCTGGATCCTTCCCG[A/C]TACAACAAGCTTATCCGCCCAGCCA
Phenotype
MIM: 118507
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
CHRNB2 PubMed Links

Gene Details

Gene
CHRNB2
Gene Name
cholinergic receptor nicotinic beta 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000748.2 369 Silent Mutation CGA,CGC R41R NP_000739.1
XM_017000180.1 369 UTR 5 XP_016855669.1

View Full Product Details