Product Details
- SNP ID
-
rs201526883
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
6
- Location
-
Chr.1:84479441 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCTGCCTTTCCGCCAGGCTTTAGC[A/G]TTTCGGAGATTAAAAACAAACAGCG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RPF1
PubMed Links
Gene Details
- Gene
- RPF1
- Gene Name
- ribosome production factor 1 homolog
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_025065.6 |
205 |
Missense Mutation |
ATT,GTT |
I54V |
NP_079341.2 |
View Full Product Details