Product Details

SNP ID
rs201593465
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:212044691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATGGAACATGTACTAGCAGTTGC[C/T]AATGAAGAAGGCTTTGTTCGATTGT
Phenotype
MIM: 610617 MIM: 611350
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DTL PubMed Links

Gene Details

Gene
DTL
Gene Name
denticleless E3 ubiquitin protein ligase homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286229.1 422 UTR 5 NP_001273158.1
NM_001286230.1 422 Silent Mutation GCC,GCT A28A NP_001273159.1
NM_016448.3 422 Silent Mutation GCC,GCT A70A NP_057532.3
XM_011509614.1 422 Silent Mutation GCC,GCT A8A XP_011507916.1
Gene
INTS7
Gene Name
integrator complex subunit 7
There are no transcripts associated with this gene.

Gene
RPL21P28
Gene Name
ribosomal protein L21 pseudogene 28
There are no transcripts associated with this gene.

View Full Product Details